Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Vejeflasker, DURAN®

20 ml, 32×50 mm
Catalog # 611-2781
Supplier:   DWK Life Sciences
Vejeflasker, DURAN®
Catalog # 611-2781
Supplier:  DWK Life Sciences
Vejeflasker, DURAN®
Vejeflasker, DURAN®

20 ml, 32×50 mm

Price
2.990,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents

DURAN® Borosilicate glass 3.3, clear.

  • Interchangeable lids with ground joint
  • Writing area in white
  • Available in low or tall form
Specifications
  • Diameter×H:
    32×50 mm
  • Kapacitet:
    20 ml
  • Seq:
    07