Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Vejeflasker, DURAN®

Vejeflasker, DURAN®

15 ml, 38×30 mm
Katalog # 611-2777
Leverandør:   DWK Life Sciences
Vejeflasker, DURAN®
Katalog # 611-2777
Leverandør:  DWK Life Sciences
Vejeflasker, DURAN®
Vejeflasker, DURAN®

15 ml, 38×30 mm

Price
3.240,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents

DURAN® Borosilicate glass 3.3, clear.

  • Interchangeable lids with ground joint
  • Writing area in white
  • Available in low or tall form
Specifikationer
  • Diameter×H:
    38×30 mm
  • Kapacitet:
    15 ml
  • Seq:
    02
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.