Orders LinkContactUsLinkComponent
Latex tests, Dryspot
Pbp2 Testkit, 50 Tests
Katalog # OXOIDR0900A
Latex tests, Dryspot
Katalog # OXOIDR0900A
CAS nummer:  
Latex tests, Dryspot
Pbp2 Testkit, 50 Tests
CAS nummer:  
Price
4.800,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
  • Simplified room temperature storage allows storage for up to two years
  • Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards
Specifikationer
Specifikationer
  • Pakn:
    50 Tests
  • Beskrivelse:
    Pbp2 Testkit