ARISTAR®, Bismuth Standard Solutions for ICP-MS
Vismut, Bi, 1000 mg/L, 2-5% HNO₃, Plastic bottle, 100 mL
Supplier: VWR Chemicals
ARISTAR®, Bismuth Standard Solutions for ICP-MS
Catalog # 456672P
Supplier: VWR Chemicals
CAS Number:
Jump to:
Product Details & Documents
- Produced from high purity acids, water ASTM I 18 Megaohms
- Solution assayed by titration
- Final concentration verified against reference material from NIST
- Total maximum uncertainty: 0,5% (typical)
- Delivered with complete certificate of analysis
: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Documents
Certificates
Search by Lot Number125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125)
Emballageoplysninger
VWR® for Calibrations
Produktlitteratur
Specifications
Specifications
- Koncentration:1000 mg/L
- Matrice:2-5% HNO₃
- Pakning:Plastic bottle
- Pakn:100 mL
- Element:Vismut
- Symbol:Bi
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



