Orders LinkContactUsLinkComponent
Bottles, wide neck, PE, rectangular, with screw cap
Bottles, wide neck, PE, rectangular, with screw cap
500 ml, 45 mm, 80×70×138 mm
Catalog # SEMA0062
Supplier:   SEMADENI
Bottles, wide neck, PE, rectangular, with screw cap
Catalog # SEMA0062
Supplier:  SEMADENI
Bottles, wide neck, PE, rectangular, with screw cap
Bottles, wide neck, PE, rectangular, with screw cap
500 ml, 45 mm, 80×70×138 mm
Price
55,30 kr
Log in for availability
1 * 1 Items
Product Details & Documents
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Kapacitet:
    500 ml
  • B×D×H:
    80×70×138 mm
  • Hals indv. Ø:
    45 mm