Orders LinkContactUsLinkComponent
Bottles, wide neck, PE, rectangular, with screw cap
Bottles, wide neck, PE, rectangular, with screw cap
100 ml, 34,5 mm, 48×42×90 mm
Katalog # SEMA0060
Leverandør:   SEMADENI
Bottles, wide neck, PE, rectangular, with screw cap
Katalog # SEMA0060
Leverandør:  SEMADENI
Bottles, wide neck, PE, rectangular, with screw cap
Bottles, wide neck, PE, rectangular, with screw cap
100 ml, 34,5 mm, 48×42×90 mm
Price
31,60 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
Specifikationer
  • Kapacitet:
    100 ml
  • B×D×H:
    48×42×90 mm
  • Hals indv. Ø:
    34,5 mm