Sample vials, trident vials, squat or tall
Squat, 7,00 ml, 23×34 mm, Yes
212-7064
: MURRAY S
Vials made from neutral glass with PE plug stoppers. Suitable for a wide variety of uses in the laboratory, production, more.
- Robust and economical, will withstand moderate temperature change
- Wide opening for easy filling and emptying
- Plug stopper for secure closure
: For production related reasons, the number of vials per box may vary.
- Kasket inkluderet:Yes
- Diameter×H:23×34 mm
- Kapacitet:7,00 ml
- Beskrivelse:Squat