Orders LinkContactUsLinkComponent
Multidishes, non treated, Nunc™
Multidishes, non treated, Nunc™
6-brønds multiskål, 3,0 ml, Pakket enkeltvis
Katalog # 735-0224
Leverandør:   Thermo Fisher Scientific
Multidishes, non treated, Nunc™
Katalog # 735-0224
Multidishes, non treated, Nunc™
Multidishes, non treated, Nunc™
6-brønds multiskål, 3,0 ml, Pakket enkeltvis
Price
2.700,00 kr
Log ind for tilgængelighed
1 * 75 Items
Product Details & Documents
PS, non treated, with lid, sterile.
  • Raised well rims to lower risk of cross contamination
  • Excellent optical quality
  • Non pyrogenic
Specifikationer
  • Pakket:
    Pakket enkeltvis
  • Anbefalet arbejdsvolumen:
    3,0 ml
  • Beskrivelse:
    6-brønds multiskål