Multidishes, non treated, Nunc™
12-brønds multiskål, 2,0 ml, Pakket enkeltvis
Katalog # 735-0223
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
PS, non treated, with lid, sterile.
- Raised well rims to lower risk of cross contamination
- Excellent optical quality
- Non pyrogenic
Specifikationer
- Pakket:Pakket enkeltvis
- Anbefalet arbejdsvolumen:2,0 ml
- Beskrivelse:12-brønds multiskål