Orders LinkContactUsLinkComponent
Adapters, reduction, Quickfit®
Adapters, reduction, Quickfit®
29/32 NS, 19/26 NS
Katalog # 201-8346
Leverandør:   SciLabware
Adapters, reduction, Quickfit®
Katalog # 201-8346
Leverandør:  SciLabware
Adapters, reduction, Quickfit®
Adapters, reduction, Quickfit®
29/32 NS, 19/26 NS
Price
638,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
PYREX®, borosilicate glass, Quickfit® joints, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
Specifikationer
  • Konusstr.:
    29/32 NS
  • Kappestr.:
    19/26 NS