Spherical ground joints, Rotulex®
joint size 13
201-8212
Made of borosilicate glass.
- All glass and PTFE pathway is highly resistant to chemical attack
- Can be used at temperatures up to 200 °C
- System comprises male ball joint with PTFE covered 'O' ring which fits into a female cup joint
- Joint can be retained together with spring loaded metal clip
Rotulex® offers a grease free spherical joint.
Bestillingsoplysninger: Component parts should be ordered separately .
- Model:joint size 13
- Seq:05