Push-in caps for tubes
LDPE, Caps, Ø×H; 8×7,5 mm, ufarvet
SEMA1445
: SEMADENI
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
- Farve:ufarvet
- Beskrivelse:Caps, Ø×H; 8×7,5 mm
- Materiale:LDPE
- Seq:07
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.