Push-in caps for tubes
LDPE, Caps, Ø×H; 7×7,5 mm, ufarvet
Katalog # SEMA1443
Leverandør: SEMADENI
Gå til:
Product Details & Documents
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
Specifikationer
- Farve:ufarvet
- Beskrivelse:Caps, Ø×H; 7×7,5 mm
- Materiale:LDPE
- Seq:06