Orders LinkContactUsLinkComponent
Flat flange lids, centre neck
Flat flange lids, centre neck
laboratorieflange DN 100, 1 hals, NS 29/32 (center), 95 mm
Catalog # LENZ06100204
Supplier:   Lenz Laborglas
Flat flange lids, centre neck
Catalog # LENZ06100204
Supplier:  Lenz Laborglas
Flat flange lids, centre neck
Flat flange lids, centre neck
laboratorieflange DN 100, 1 hals, NS 29/32 (center), 95 mm
Price
1.630,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
Documents
Specifications
  • Model:
    1 hals
  • med:
    laboratorieflange DN 100
  • Hals:
    NS 29/32 (center)
  • Samlet højde:
    95 mm