Acetonitril ≥99%, TECHNICAL
Metal drum, 200 L
Leverandør: VWR Chemicals
Acetonitril ≥99%, TECHNICAL
Katalog # 20070.553
Leverandør: VWR Chemicals
CAS nummer: 75-05-8
Gå til:
Product Details & Documents
Synonymer: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- CAS nummer:75-05-8
- Formel:H₃CCN
- MDL nummer:MFCD00001878
- Molecular Weight:41.05 g/mol
- Merck Index:13,00071
- Kogende Pt:81,6 °C (1013 hPa)
- Smeltende Pt:–45,7 °C
- Tæthed:0,782 g/cm³ (20 °C)
- Flash Pt:2 °C (closed cup)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-835-2
- FN:1648
- ADR:3,II
- NÅ:01-2119471307-38
Documents
Certifikater
Søg efter partinummer1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125)
Emballageoplysninger
P20_VWRC20070.291_SPEC_EN.pdf P20_VWRC20070.325_SPEC_EN.pdf P20_VWRC20070.462_SPEC_EN.pdf P20_VWRC20070.553_SPEC_EN.pdf P20_VWRC20070.950_SPEC_EN.pdf
Produktspecifikation
More
VWR Chemicals Big Packs and Bulks
Produktlitteratur
Specifikationer
Specifikationstestresultater
- Assay:Min. 99 %
- Refractive index (20°C):1.343 - 1.345
Specifikationer
- Pakning:Metal drum
- Pakn:200 L
- Cat. No.:20070.553
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø