Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

X-Gal

Glass bottle for solids, 100 mg
Catalog # 10011.
Supplier:   Biotium
X-Gal
Catalog # 10011.
Supplier:  Biotium
CAS Number:  7240-90-6
X-Gal

Glass bottle for solids, 100 mg

CAS Number:  7240-90-6
Price
599,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
  • CAS Number:7240-90-6
  • Formula:C₁₄H₁₅BrClNO₆
  • MDL Number:MFCD00005666
  • Molecular Weight:408.63 g/mol
  • Melting Pt:237…239 °C
  • Storage Temperature:Fryser
  • EINECS:230-640-8
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 mg

Specifications