Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
X-Gal

X-Gal

Glass bottle for solids, 5 g
Catalog # 10011-2
Supplier:   Biotium
X-Gal
Catalog # 10011-2
Supplier:  Biotium
CAS Number:  7240-90-6
X-Gal

Glass bottle for solids, 5 g

CAS Number:  7240-90-6
Price
6.330,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
  • CAS nummer:7240-90-6
  • Formel:C₁₄H₁₅BrClNO₆
  • MDL nummer:MFCD00005666
  • Molecular Weight:408.63 g/mol
  • Smeltende Pt:237…239 °C
  • Opbevaringstemperatur:Fryser
  • EINECS:230-640-8

Documents

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer