Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Beakers, low form, Azlon®

50 ml, 60 mm, 43 mm
  213-0139
 :   SciLabware
Beakers, low form, Azlon®
  213-0139
Beakers, low form, Azlon®
Beakers, low form, Azlon®

50 ml, 60 mm, 43 mm

 
200,00 kr
   
1 * 1 Items
 

PTFE, opaque, rigid, non-porous.

  • Excellent chemical resistance - resists nearly all chemicals except fluorine and molten alkali metals
  • Can withstand 260 °C
  • Effective substitutes for platinum containers

 : It is NOT advisable to use PTFE beakers on a hotplate.

 
  • Udv.Ø:
    43 mm
  • Kapacitet:
    50 ml
  • Højde:
    60 mm