Orders LinkContactUsLinkComponent
Bottle top tilt measures
Bottle top tilt measures
50 ml, klar
Katalog # 613-6255
Leverandør:   Lenz Laborglas
Bottle top tilt measures
Katalog # 613-6255
Leverandør:  Lenz Laborglas
Bottle top tilt measures
Bottle top tilt measures
50 ml, klar
Price
700,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This dispenser head is completed by a 1 litre bottle for repetitive dispensing of the same volume.
NS 29/32
Documents
Specifikationer
  • Kapacitet:
    50 ml
  • Farve:
    klar