Bottle top tilt measures
50 ml, klar
Katalog # 613-6255
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
This dispenser head is completed by a 1 litre bottle for repetitive dispensing of the same volume.
NS 29/32
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Kapacitet:50 ml
- Farve:klar