Incinerating dishes, rectagular
40 mm, 8,5 mm, 10,5 mm
Katalog # HALDALGK40X10X8
Leverandør: MORGAN ADVANCED MATERIALS HALD
Incinerating dishes, rectagular
Katalog # HALDALGK40X10X8
Leverandør: MORGAN ADVANCED MATERIALS HALD
Gå til:
Product Details & Documents
Alsint 99,7.
- Good thermal shock resistance due to high thermal conductivity
- Resistant to chemical attack, e.g. hydrogen and other reducing gases
Specifikationer
- Bredde:10,5 mm
- Højde:8,5 mm
- Længde:40 mm
- Seq:01