Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Incinerating dishes, rectagular

150 mm, 35 mm, 65 mm
  HALDALGK150X65X35
Incinerating dishes, rectagular
  HALDALGK150X65X35
Incinerating dishes, rectagular
Incinerating dishes, rectagular

150 mm, 35 mm, 65 mm

 
4.250,00 kr
   
1 * 1 Items
 

Alsint 99,7.

  • Good thermal shock resistance due to high thermal conductivity
  • Resistant to chemical attack, e.g. hydrogen and other reducing gases
 
  • Bredde:
    65 mm
  • Højde:
    35 mm
  • Længde:
    150 mm
  • Seq:
    13