Temperature stabilisation jacket
Temperature stabilisation jacket with support plate, 577 00
Katalog # SCOI285405508
Leverandør: SI Analytics
Gå til:
Product Details & Documents
The temperature of capillary viscometers can be stabilised in this jacket using circulation thermostats from 0 to 180 °C.
- Support plate to facilitate the use
- Quick-action seal to simplify the changing viscometers
- Additional hole in the support plate so that a control thermometer can fit
Suitable for use with Ubbelohde viscometers.
Specifikationer
- Model:577 00
- Beskrivelse:Temperature stabilisation jacket with support plate
- Seq:001