Screw Caps for Bottles
Screw caps with foamed PE seal, 80 mm, rød, PP
Katalog # RIXI8-2001-80-R
Leverandør: BERLIN PACKAGING RIXIUS GMBH
Gå til:
Product Details & Documents
Specifikationer
- Gevind:80 mm
- Farve:rød
- Beskrivelse:Screw caps with foamed PE seal
- Materiale:PP
- Seq:43