Multidishes, non treated, Nunc™
24-brønds multiskål, 1,0 ml, Pakket enkeltvis
734-0994
PS, non treated, with lid, sterile.
- Raised well rims to lower risk of cross contamination
- Excellent optical quality
- Non pyrogenic
- Pakket:Pakket enkeltvis
- Anbefalet arbejdsvolumen:1,0 ml
- Beskrivelse:24-brønds multiskål