Orders LinkContactUsLinkComponent
Multidishes, non treated, Nunc™
24-brønds multiskål, 1,0 ml, Pakket enkeltvis
  734-0994
Multidishes, non treated, Nunc™
  734-0994
Multidishes, non treated, Nunc™
Multidishes, non treated, Nunc™
24-brønds multiskål, 1,0 ml, Pakket enkeltvis
2.900,00 kr
1 * 75 Items
PS, non treated, with lid, sterile.
  • Raised well rims to lower risk of cross contamination
  • Excellent optical quality
  • Non pyrogenic
  • Pakket:
    Pakket enkeltvis
  • Anbefalet arbejdsvolumen:
    1,0 ml
  • Beskrivelse:
    24-brønds multiskål