Acetonitril
Glass bottle, 1 L
Catalog # ROTH8825.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- CAS Number:75-05-8
- Formula:H₃CCN
- MDL Number:MFCD00001878
- Molecular Weight:41.05 g/mol
- Merck Index:13,00071
- Boiling Pt:81,6 °C (1013 hPa)
- Melting Pt:–45,7 °C
- Density:0,782 g/cm³ (20 °C)
- Flash Pt:2 °C (closed cup)
- Storage Temperature:Stue temperatur
- EINECS:200-835-2
- UN:1648
- ADR:3,II
- REACH:01-2119471307-38
Specifications
Specifications
- Pakning:Glass bottle
- Pakn:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



