Volumetric Flasks, Class A
500 ml, 19/26, 0,25 ± ml
Katalog # WITG3.664.050.BL
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
Specifikationer
- NS:19/26
- Kapacitet:500 ml
- Seq:13
- Tolerance:0,25 ± ml