Orders LinkContactUsLinkComponent
Volumetric Flasks, Class A
Volumetric Flasks, Class A
50 ml, 14/23, 0,10 ± ml
Catalog # WITG3.664.006.BL
Supplier:   witeg Labortechnik
Volumetric Flasks, Class A
Catalog # WITG3.664.006.BL
Supplier:  witeg Labortechnik
Volumetric Flasks, Class A
Volumetric Flasks, Class A
50 ml, 14/23, 0,10 ± ml
Price
245,00 kr
Log in for availability
1 * 2 Items
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
  • DE-M marked according to the German calibration law
  • Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
Specifications
  • NS:
    14/23
  • Kapacitet:
    50 ml
  • Seq:
    9
  • Tolerance:
    0,10 ± ml