Orders LinkContactUsLinkComponent
Sulfonation flasks, with three necks
Sulfonation flasks, with three necks
100 ml, 29/32 NS, 14/23 NS
Katalog # WITG0.851.101
Leverandør:   witeg Labortechnik
Sulfonation flasks, with three necks
Katalog # WITG0.851.101
Leverandør:  witeg Labortechnik
Sulfonation flasks, with three necks
Sulfonation flasks, with three necks
100 ml, 29/32 NS, 14/23 NS
Price
1.210,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one parallel and two angled side necks.
Specifikationer
  • Kapacitet:
    100 ml
  • Seq:
    01
  • Sidehalse:
    14/23 NS
  • Centreret hals:
    29/32 NS