Sulfonation flasks, with three necks
200 ml, 29/32 NS, 14/23 NS
Catalog # WITG0.851.020
Supplier: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With one parallel and two angled side necks.
Specifikationer
- Kapacitet:200 ml
- Seq:02
- Sidehalse:14/23 NS
- Centreret hals:29/32 NS