Glass stoppers with hexagonal grip
19/26, Klart glas
WITG0.302.019
Hollow standard taper stoppers with hexagonal grip, made of borosilicate glass 3.3.
- Acc. to DIN 12252
- NS:19/26
- Model:Klart glas
Hollow standard taper stoppers with hexagonal grip, made of borosilicate glass 3.3.