Orders LinkContactUsLinkComponent
Glass stoppers with hexagonal grip
19/26, Klart glas
  WITG0.302.019
Glass stoppers with hexagonal grip
  WITG0.302.019
Glass stoppers with hexagonal grip
Glass stoppers with hexagonal grip
19/26, Klart glas
28,10 kr
1 * 1 Items
Hollow standard taper stoppers with hexagonal grip, made of borosilicate glass 3.3.
  • Acc. to DIN 12252
  • NS:
    19/26
  • Model:
    Klart glas