Orders LinkContactUsLinkComponent
Screwthread joints for VACUU.LAN®
Straight adapter, for G 3/8" thread to PTFE tubing 10/8 mm
  VACU20639855
 :   VACUUBRAND
Screwthread joints for VACUU.LAN®
  VACU20639855
Screwthread joints for VACUU.LAN®
Straight adapter, for G 3/8" thread to PTFE tubing 10/8 mm
176,00 kr
1 * 1 Items
Adapters for VACUU.LAN® local area vacuum network components.
  • Beskrivelse:
    Straight adapter, for G 3/8" thread to PTFE tubing 10/8 mm
  • Seq:
    01