Screwthread joints for VACUU.LAN®
Elbow (90°) adapter, 28 mm, for 1/8" NPT thread to PTFE tubing 10/8 mm
Katalog # VACU20639853
Leverandør: VACUUBRAND
Gå til:
Product Details & Documents
Adapters for VACUU.LAN® local area vacuum network components.
Specifikationer
- Beskrivelse:Elbow (90°) adapter, 28 mm, for 1/8" NPT thread to PTFE tubing 10/8 mm
- Seq:03
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



