Orders LinkContactUsLinkComponent
Funnels with short stem
Funnels with short stem
150 mm, 40 mm, 400 mm
Katalog # SEMA0202
Leverandør:   SEMADENI
Funnels with short stem
Katalog # SEMA0202
Leverandør:  SEMADENI
Funnels with short stem
Funnels with short stem
150 mm, 40 mm, 400 mm
Price
458,00 kr
Log ind for tilgængelighed
1 * 1 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Funnels, made of HDPE, with smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
  • Easy to clean
  • Good chemical resistance
  • Good temperature resistance
Specifikationer
  • Tragt udv. Ø:
    400 mm
  • Stilk udv. Ø:
    40 mm
  • Stilklængde:
    150 mm