Orders LinkContactUsLinkComponent
Multidishes, non treated, Nunc™
Multidishes, non treated, Nunc™
4-brønds multiskål, 1,0 ml, 4/pose
Katalog # 734-2375
   Lav fremstillingspåvirkning
Leverandør:   Thermo Fisher Scientific
Multidishes, non treated, Nunc™
Katalog # 734-2375
   Lav fremstillingspåvirkning
Multidishes, non treated, Nunc™
Multidishes, non treated, Nunc™
4-brønds multiskål, 1,0 ml, 4/pose
Price
3.230,00 kr
Log ind for tilgængelighed
1 * 120 Items
Product Details & Documents
PS, non treated, with lid, sterile.
  • Raised well rims to lower risk of cross contamination
  • Excellent optical quality
  • Non pyrogenic
Specifikationer
  • Pakket:
    4/pose
  • Anbefalet arbejdsvolumen:
    1,0 ml
  • Beskrivelse:
    4-brønds multiskål
  • Cat. No.:
    734-2375
  • Miljøcertificering:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Lav fremstillingspåvirkning