Multidishes, non treated, Nunc™
4-brønds multiskål, 1,0 ml, 4/pose
Katalog # 734-2375
Leverandør: Thermo Fisher Scientific
Multidishes, non treated, Nunc™
Katalog # 734-2375
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
PS, non treated, with lid, sterile.
- Raised well rims to lower risk of cross contamination
- Excellent optical quality
- Non pyrogenic
Specifikationer
- Pakket:4/pose
- Anbefalet arbejdsvolumen:1,0 ml
- Beskrivelse:4-brønds multiskål
- Cat. No.:734-2375
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning