Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Centrifugerør, ASTM

Centrifugerør, ASTM

Cylindrisk, konisk bund, ASTM D91, 100 ml, 203 mm, uden cap
Katalog # LENZ05675000
Leverandør:   Lenz Laborglas
Centrifugerør, ASTM
Katalog # LENZ05675000
Leverandør:  Lenz Laborglas
Centrifugerør, ASTM
Centrifugerør, ASTM

Cylindrisk, konisk bund, ASTM D91, 100 ml, 203 mm, uden cap

Price
368,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

DURAN®, borosilicate glass.

  • Withstands forces up to 700 ×g
  • Good temperature resistance
  • Good chemical resistance

With conical bottom or pear shaped.

Documents

Specifikationer
  • Lukkestørrelse:
    uden cap
  • Kapacitet:
    100 ml
  • Beskrivelse:
    Cylindrisk, konisk bund, ASTM D91
  • Længde:
    203 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.