Orders LinkContactUsLinkComponent
Pulverkolber
1000 ml, 29/32 NS
Katalog # 201-0628
Leverandør:   Lenz Laborglas
Pulverkolber
Katalog # 201-0628
Leverandør:  Lenz Laborglas
Pulverkolber
Pulverkolber
1000 ml, 29/32 NS
Price
1.160,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for rotary evaporators, for drying of powder-like substances. With center neck, conical joint and inscription label.
Documents
Specifikationer
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    1000 ml