Orders LinkContactUsLinkComponent
Cover glasses
24×50 mm, #1
Katalog # KNITVD12450Y100A
Leverandør:   KNITTEL
Cover glasses
Katalog # KNITVD12450Y100A
Leverandør:  KNITTEL
Cover glasses
24×50 mm, #1
Price
863,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
 : ISO 8255/1
 : 100 pieces per box - 1000 pieces per carton.
Specifikationer
  • Antal:
    #1
  • L×B:
    24×50 mm