Cover glasses
24×50 mm, #1
Katalog # KNITVD12450Y100A
Leverandør: KNITTEL
Gå til:
Product Details & Documents
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
: ISO 8255/1
: 100 pieces per box - 1000 pieces per carton.
Specifikationer
- Antal:#1
- L×B:24×50 mm