Orders LinkContactUsLinkComponent
Cover glasses
Cover glasses
Cover glasses, square, without cords and blisters, 24×24 mm, #1
Katalog # KNITVD12424Y100A
Cover glasses
Katalog # KNITVD12424Y100A
Leverandør:  
Cover glasses
Cover glasses, square, without cords and blisters, 24×24 mm, #1
Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
COVER GLASS 24X24MM, NR.1 1 * 1.000 Items Katalog #631-1571
Product 631-1571 is similar to KNITVD12424Y100A but manufacturer and specifications may vary.
Product Details & Documents
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
Certifications: ISO 8255/1
Packaging: 100 pieces per box - 1000 pieces per carton.
Documents
Specifikationer
  • Antal:
    #1
  • Beskrivelse:
    Cover glasses, square, without cords and blisters
  • L×B:
    24×24 mm