Cover glasses
18×18 mm, 0,16 mm
KNITVD11818Y100A
: KNITTEL
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
: ISO 8255/1
: 100 pieces per box - 1000 pieces per carton.
- L×B:18×18 mm
- Tykkelse:0,16 mm
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
: ISO 8255/1
: 100 pieces per box - 1000 pieces per carton.