Orders LinkContactUsLinkComponent
Cover glasses
18×18 mm, 0,16 mm
  KNITVD11818Y100A
 :   KNITTEL
Cover glasses
  KNITVD11818Y100A
 :  KNITTEL
Cover glasses
18×18 mm, 0,16 mm
523,00 kr
1 * 1.000 Items
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
 : ISO 8255/1
 : 100 pieces per box - 1000 pieces per carton.
  • L×B:
    18×18 mm
  • Tykkelse:
    0,16 mm