Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Conductivity standards, Certipur® Supelco®

Conductivity standards, Certipur® Supelco®

KCl 0,001 mol/l, 147 µS/cm, sachets of 30 ml, Sachet, 30
Katalog # 1.01586.0001
Leverandør:   Merck
Conductivity standards, Certipur® Supelco®
Katalog # 1.01586.0001
Leverandør:  Merck
CAS nummer:  
Conductivity standards, Certipur® Supelco®
Conductivity standards, Certipur® Supelco®

KCl 0,001 mol/l, 147 µS/cm, sachets of 30 ml, Sachet, 30

CAS nummer:  
Price
840,00 kr
Log ind for tilgængelighed
1 * 30 Items
Product Details & Documents

These conductivity sachets are convenient, easy to use and ensure accurate and reliable results both in the field or in the laboratory. The 30 ml sachets always provide a fresh solution preventing contamination by microorganisms, CO₂ or other substances. A conductivity sensor can be simply inserted into the sachet.

  • Traceable to PTB and NIST
  • Tested against primary reference standards

Certipur conductivity standards are also available in 500 ml packs for the user with higher volume requirements.

Specifikationer

Specifikationer

  • Pakning:
    Sachet
  • Pakn:
    30
  • Beskrivelse:
    KCl 0,001 mol/l, 147 µS/cm, sachets of 30 ml
  • Cat. No.:
    1.01586.0001
  • M-Clarity quality level:
    MQ100

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.