Probes for precision digital thermometer
Surface Probe, ad 20 th, ±0,3 °C or 0,5% of range, 8,0×105 mm
AMARE910656
: Amarell
Precise Pt 100 class A probe of stainless steel, PVC-cable.
- Beskrivelse:Surface Probe, ad 20 th
- Seq:04
- Nøjagtighed:±0,3 °C or 0,5% of range
- Ø×L:8,0×105 mm



