Orders LinkContactUsLinkComponent
Glass beads
Glass beads
Glass beads, solid, 1 mm
Katalog # 201-0276
Glass beads
Katalog # 201-0276
Glass beads
Glass beads
Glass beads, solid, 1 mm
Price
172,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Solid glass.
For packing distillation columns.
Specifikationer
  • Ø:
    1 mm
  • Beskrivelse:
    Glass beads, solid
  • Seq:
    01