EDTA, Supelco®
Plastic bottle, 250 g
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
- CAS nummer:60-00-4
- Formel:C₁₀H₁₆N₂O₈
- MDL nummer:MFCD00003541
- Molecular Weight:292.25 g/mol
- Merck Index:13,03546
- Kogende Pt:614 °C (1013 hPa)
- Smeltende Pt:220 °C
- Tæthed:0,86 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:>100 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-449-4
- NÅ:01-2119486399-18
Documents
Specifikationer
Specifikationstestresultater
- Solubility:0.5 g/l (20 °C)
- Melting Point:220 °C (decomposition)
- Molar Mass:292.24 g/mol
- Bulk Density:600 kg/m3
- Vapor Pressure:- 0.013 hPa (20 °C)
- Flash Point:100 °C DIN 51758
- Density:0.86 g/cm3 (20 °C)
- pH:2.5 (10 g/l, H2O, 23 °C) (slurry)
- Ignition Point:200 °C
Specifikationer
- Pakning:Plastic bottle
- Pakn:250 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



