Orders LinkContactUsLinkComponent
Stirring shafts
Stirring shafts
320 mm, 7 mm
Catalog # RSGA607.320
Supplier:   RSG
Stirring shafts
Catalog # RSGA607.320
Supplier:  RSG
Stirring shafts
Stirring shafts
320 mm, 7 mm
Price
122,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Ø:
    7 mm
  • Længde:
    320 mm
  • Seq:
    007