Pharmacist's spatulas
Flexible blade, 75 mm, 18/22 mm
Catalog # 231-0547
Supplier: RSG
Jump to:
Product Details & Documents
Stainless steel blade, wooden handle.
- Available with flexible blade (Remanit 4310) or with hardened blade (INOX 4034)
- 75 to 150 mm: conical blade
Specifications
- Beskrivelse:Flexible blade
- Bladbredde:18/22 mm
- Klinge længde:75 mm