Apoteksspatler
Fleksibelt blad, 75 mm, 18/22 mm
Katalog # RSGA051.075
Leverandør: RSG
Gå til:
Product Details & Documents
Pharmacist's spatulas with stainless steel blade and black plastic handle.
- Conical blade
- Dishwasher-proof
- Available flexible or non flexible
Specifikationer
- Beskrivelse:Fleksibelt blad
- Bladbredde:18/22 mm
- Klinge længde:75 mm