Orders LinkContactUsLinkComponent
Forceps, straight
Forceps, straight
Fine tips, 130 mm
Katalog # 232-0074
Leverandør:   RSG
Forceps, straight
Katalog # 232-0074
Leverandør:  RSG
Forceps, straight
Forceps, straight
Fine tips, 130 mm
Price
437,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic.
  • Serrated tips
  • With guide pin
Specifikationer
  • Model:
    Fine tips
  • Længde:
    130 mm