Pharmacist's spatulas
Hardened blade, 100 mm, 18/22 mm
Katalog # 231-0552
Leverandør: RSG
Gå til:
Product Details & Documents
Stainless steel blade, wooden handle.
- Available with flexible blade (Remanit 4310) or with hardened blade (INOX 4034)
- 75 to 150 mm: conical blade
Specifikationer
- Beskrivelse:Hardened blade
- Bladbredde:18/22 mm
- Klinge længde:100 mm