Orders LinkContactUsLinkComponent
Pharmacist's spatulas
Pharmacist's spatulas
Hardened blade, 150 mm, 18/22 mm
Katalog # 231-0554
Leverandør:   RSG
Pharmacist's spatulas
Katalog # 231-0554
Leverandør:  RSG
Pharmacist's spatulas
Pharmacist's spatulas
Hardened blade, 150 mm, 18/22 mm
Price
930,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Stainless steel blade, wooden handle.
  • Available with flexible blade (Remanit 4310) or with hardened blade (INOX 4034)
  • 75 to 150 mm: conical blade
Specifikationer
  • Beskrivelse:
    Hardened blade
  • Bladbredde:
    18/22 mm
  • Klinge længde:
    150 mm