Instrument trays, rectangular
20 mm, 110 mm, 170 mm
Katalog # RSGA701.217
Leverandør: RSG
Gå til:
Product Details & Documents
Stainless steel, 18/10.
- Smooth surfaces for fast, easy cleaning
- Good chemical resistance
- Very good thermal resistance
Documents
RSGA701.242_TDS_EN
Datablad
Specifikationer
- Bredde:110 mm
- Højde:20 mm
- Længde:170 mm